Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001283061.1 | NP_001269990.1 | 13 | 1384 | |
NM_001283062.1 | NP_001269991.1 | 11 | 1270 | ||
NM_007968.3 | NP_031994.2 | 13 | 1381 | ||
GenBank mRNA | AK014366.1 | - | 5 | 350 | |
AK019460.1 | - | 13 | 1330 | ||
AK034755.1 | - | 13 | 1357 | ||
AK049743.1 | - | 11 | 1250 | ||
AK147103.1 | - | 13 | 1348 | ||
AK150345.1 | - | 13 | 1342 | ||
AK151625.1 | - | 13 | 1352 | ||
AK153276.1 | - | 13 | 1342 | ||
AK200262.1 | - | 5 | 354 | ||
BC068226.1 | - | 13 | 1351 | 5707017 | |
X79233.1 | - | 13 | 1370 |