Gene Transcripts

Gene Symbol
Gpi1
Entrez Gene ID
14751
Gene Name
glucose phosphate isomerase 1
Gene Aliases
Amf, Gpi, Gpi-1, Gpi-1r, Gpi-1s, Gpi-1t, Gpi1-r, Gpi1-s, Gpi1-t, Gpi1s, MF, NK, NK/GPI, Nlk, Org, Pgi, Phi
Chromosome Location
Chr. 7: 34201327 - 34230336 on Build GRCm38
UniGene ID
Mm.589
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008155.4 NP_032181.2 2 317
GenBank mRNA AK031081.1 - 2 222
AK133827.1 - 2 239
AK134890.1 - 2 251
AK137805.1 - 2 245
AK147124.1 - 2 280
AK150341.1 - 2 248
AK159838.1 - 2 221
AK164566.1 - 2 216
AK169681.1 - 2 246
AK171956.1 - 1 938
BC086640.1 - 2 264 6832250
BC088995.1 - 2 224 6403790
BC157972.1 - 2 230 9007294
BC172124.1 - 2 230 9054410
BY287889.1 - 2 287
BY350616.1 - 2 317
M14220.1 - 2 246

View Full Product Details