Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008325.4 | NP_032351.2 | 4 | 745 | |
NM_172997.1 | NP_766585.1 | 3 | 604 | ||
GenBank mRNA | AK031414.1 | - | 3 | 399 | |
AK045330.1 | - | 2 | 232 | ||
AK155627.1 | - | 4 | 746 | ||
AK169998.1 | - | 3 | 403 | ||
BC057935.1 | - | 3 | 366 | 5327150 | |
L34111.1 | - | 3 | 319 |