Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008702.3 | NP_032728.3 | 9, 10 | 2595 | |
GenBank mRNA | AF036332.1 | - | 9, 10 | 1722 | |
AK078118.1 | - | 9, 10 | 1704 | ||
BC057667.1 | - | 9, 10 | 1680 | 4235238 | |
BC058652.1 | - | 9, 10 | 2543 | 6406851 |