Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008812.2 | NP_032838.2 | 2, 3 | 382 | |
GenBank mRNA | AK148156.1 | - | 2, 3 | 369 | |
AK154814.1 | - | 2, 3 | 368 | ||
AK158382.1 | - | 2, 3 | 369 | ||
AK171275.1 | - | 2, 3 | 369 | ||
BB850871.1 | - | 2, 3 | 384 | ||
BC040350.1 | - | 2, 3 | 324 | 4192742 | |
BC049947.1 | - | 2, 3 | 355 | 3600978 | |
D16580.1 | - | 2, 3 | 315 |