Gene Transcripts

Gene Symbol
Ppl
Entrez Gene ID
19041
Gene Name
periplakin
Gene Aliases
AW553870
Chromosome Location
Chr. 16: 5086291 - 5132481 on Build GRCm38
UniGene ID
Mm.266875
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008909.2 NP_032935.2 19, 20 2537
GenBank mRNA AF126834.1 - 19, 20 2477

View Full Product Details