Gene Transcripts
- Gene Symbol
- BANF1
- Entrez Gene ID
- 8815
- Gene Name
- barrier to autointegration factor 1
- Gene Aliases
- BAF, BCRP1, D14S1460, NGPS
- Chromosome Location
- Chr. 11: 66002079 - 66004149 on Build GRCh38
- UniGene ID
- Hs.433759
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001143985.1 |
NP_001137457.1
|
2, 3 |
582 |
|
NM_003860.3 |
NP_003851.1
|
2, 3 |
639 |
|
XM_017018514.1 |
XP_016874003.1
|
2, 3 |
632 |
|
XM_017018515.1 |
XP_016874004.1
|
2, 3 |
172 |
|
GenBank mRNA |
AF044773.1 |
- |
2, 3 |
235 |
|
AF068235.1 |
- |
2, 3 |
638 |
|
AF070447.1 |
- |
1, 2 |
131 |
|
AK311961.1 |
- |
2, 3 |
206 |
|
BC005942.1 |
- |
2, 3 |
218 |
4074168 |
BC107702.1 |
- |
2, 3 |
315 |
6495215 |
BG748852.1 |
- |
2, 3 |
422 |
|
BU940708.1 |
- |
2, 3 |
399 |
|
CR542140.1 |
- |
1, 2 |
131 |
|
View Full Product Details