Gene Transcripts

Gene Symbol
Scg2
Entrez Gene ID
20254
Gene Name
secretogranin II
Gene Aliases
Chgc, SgII
Chromosome Location
Chr. 1: 79434669 - 79440121 on Build GRCm38
UniGene ID
Mm.5038
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009129.2 NP_033155.1 2 468
GenBank mRNA BC014717.1 - 2 437
BC048249.1 - 2 368 6493827
BI735349.1 - 2 439
CK619591.1 - 2 468

View Full Product Details