Gene Transcripts

Gene Symbol
C3
Entrez Gene ID
12266
Gene Name
complement component 3
Gene Aliases
AI255234, ASP, HSE-MSF, Plp
Chromosome Location
Chr. 17: 57203968 - 57228136 on Build GRCm38
UniGene ID
Mm.19131
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC043338.1 - 6, 7 722
K02782.1 - 6, 7 736
M35659.1 - 5, 6 608

View Full Product Details