Gene Transcripts

Gene Symbol
Chrd
Entrez Gene ID
12667
Gene Name
chordin
Gene Aliases
Chd
Chromosome Location
Chr. 16: 20731417 - 20742384 on Build GRCm38
UniGene ID
Mm.20457
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278041.1 NP_001264970.1 3, 4 476
NM_009893.2 NP_034023.1 3, 4 410
GenBank mRNA AF069501.1 - 3, 4 410
AK077460.1 - 3, 4 404
AK149445.1 - 3, 4 385
AK165339.1 - 3, 4 404
BC145017.1 - 3, 4 474
BC145018.1 - 3, 4 474
BC145019.1 - 3, 4 408
BC145696.1 - 3, 4 474 40130999

View Full Product Details