Gene Transcripts

Gene Symbol
Dyrk1b
Entrez Gene ID
13549
Gene Name
dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 1b
Gene Aliases
Mirk
Chromosome Location
Chr. 7: 28179483 - 28187793 on Build GRCm38
UniGene ID
Mm.57249
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037957.3 NP_001033046.1 3, 4 450
NM_001271370.1 NP_001258299.1 3, 4 403
NM_010092.2 NP_034222.1 3, 4 450
GenBank mRNA AJ537610.1 - 3, 4 403
AK148342.1 - 3, 4 430
BC019545.1 - 3, 4 450 4511845
Y18280.2 - 3, 4 439

View Full Product Details