Gene Transcripts

Gene Symbol
Dpm1
Entrez Gene ID
13480
Gene Name
dolichol-phosphate (beta-D) mannosyltransferase 1
Gene Aliases
AI118379, AI194292
Chromosome Location
Chr. 2: 168209048 - 168230379 on Build GRCm38
UniGene ID
Mm.422657
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010072.3 NP_034202.1 9 1175
GenBank mRNA AK004834.1 - 9 1168
AK005268.1 - 9 1161

View Full Product Details