Gene Transcripts
- Gene Symbol
- Htr5b
- Entrez Gene ID
- 15564
- Gene Name
- 5-hydroxytryptamine (serotonin) receptor 5B
- Gene Aliases
- 5-Ht5b
- Chromosome Location
- Chr. 1: 121509758 - 121529312 on Build GRCm38
- UniGene ID
- Mm.4833
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010483.3 |
NP_034613.2
|
2 |
1552 |
|
GenBank mRNA |
X69867.1 |
- |
2 |
1587 |
|
View Full Product Details