Gene Transcripts

Gene Symbol
Hoxc6
Entrez Gene ID
15425
Gene Name
homeobox C6
Gene Aliases
Hox-3.3, Hox-6.1
Chromosome Location
Chr. 15: 102998226 - 103011882 on Build GRCm38
UniGene ID
Mm.4444
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010465.2 NP_034595.2 2 1442
GenBank mRNA AK050798.1 - 2 1210
X16510.1 - 1418
X16511.1 - 4 1540

View Full Product Details