Gene Transcripts
- Gene Symbol
- Itga2b
- Entrez Gene ID
- 16399
- Gene Name
- integrin alpha 2b
- Gene Aliases
- AI172977, CD41, CD41B, GpIIb, alphaIIb
- Chromosome Location
- Chr. 11: 102453297 - 102469883 on Build GRCm38
- UniGene ID
- Mm.26646
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010575.2 |
NP_034705.2
|
1, 2 |
353 |
|
GenBank mRNA |
AF166384.1 |
- |
1, 2 |
231 |
|
AF170316.1 |
- |
1, 2 |
214 |
|
AK142289.1 |
- |
1, 2 |
215 |
|
AK154619.1 |
- |
1, 2 |
354 |
|
AK156872.1 |
- |
1, 2 |
216 |
|
AK157012.1 |
- |
1, 2 |
215 |
|
AK158443.1 |
- |
1, 2 |
200 |
|
BC120493.1 |
- |
1, 2 |
203 |
40129416 |
BC137570.1 |
- |
1, 2 |
203 |
8860589 |
BY204133.1 |
- |
1, 2 |
354 |
|
View Full Product Details