Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163569.1 | NP_001157041.1 | 3 | 332 | |
NM_010628.3 | NP_034758.2 | 3 | 337 | ||
GenBank mRNA | AJ132889.1 | - | 3 | 270 | |
AK029506.1 | - | 3 | 230 | ||
AK029776.1 | - | 3 | 302 | ||
AK087008.1 | - | 3 | 302 | ||
BC115765.1 | - | 4 | 530 | 40059805 | |
BY108772.1 | - | 3 | 333 | ||
CJ139757.1 | - | 2 | 240 |