Gene Transcripts

Gene Symbol
Krtap13
Entrez Gene ID
16699
Gene Name
keratin associated protein 13
Gene Aliases
4C32
Chromosome Location
Chr. 16: 88750744 - 88751628 on Build GRCm38
UniGene ID
Mm.353193
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010671.1 NP_034801.1 1 493
GenBank mRNA AF031485.1 - 1 493
AK003960.1 - 1 516
BC063754.1 - 1 500 30293948

View Full Product Details