Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010686.3 | NP_034816.1 | 8 | 1130 | |
GenBank mRNA | AK031802.1 | - | 8 | 1144 | |
AK050633.1 | - | 8 | 1145 | ||
AK089432.1 | - | 8 | 1180 | ||
AK089508.1 | - | 8 | 1153 | ||
AK089535.1 | - | 8 | 1145 | ||
AK150299.1 | - | 8 | 1142 | ||
AK152918.1 | - | 8 | 1141 | ||
AK153877.1 | - | 8 | 1144 | ||
AK155353.1 | - | 3 | 587 | ||
AK155744.1 | - | 8 | 1140 | ||
AK163146.1 | - | 8 | 1145 | ||
AK170438.1 | - | 8 | 1132 | ||
AK170690.1 | - | 9 | 1296 | ||
AK170867.1 | - | 8 | 1148 | ||
AK171053.1 | - | 8 | 1155 | ||
AK171057.1 | - | 8 | 1181 | ||
AK171105.1 | - | 8 | 1146 | ||
AK171116.1 | - | 8 | 1145 | ||
AK171149.1 | - | 8 | 1153 | ||
AK171172.1 | - | 1145 | |||
AK171193.1 | - | 8 | 1145 | ||
AK171213.1 | - | 8 | 1145 | ||
AK171236.1 | - | 8 | 1145 | ||
AK171244.1 | - | 8 | 1145 | ||
AK171250.1 | - | 8 | 1166 | ||
AK171269.1 | - | 8 | 1182 | ||
AK171270.1 | - | 8 | 1148 | ||
AK171274.1 | - | 8 | 1138 | ||
AK171280.1 | - | 8 | 1178 | ||
AK171285.1 | - | 8 | 1182 | ||
AK171481.1 | - | 8 | 1181 | ||
AK171547.1 | - | 8 | 1145 | ||
AK171562.1 | - | 8 | 1138 | ||
AK171571.1 | - | 8 | 1164 | ||
AK171575.1 | - | 8 | 1182 | ||
AK172306.1 | - | 8 | 1148 | ||
AK172486.1 | - | 8 | 1180 | ||
AK172707.1 | - | 8 | 1145 | ||
AK172717.1 | - | 5 | 850 | ||
AK172885.1 | - | 8 | 1120 | ||
BC020993.1 | - | 8 | 1130 | 4037686 | |
BC055057.1 | - | 8 | 1117 | 6529760 | |
BC158032.1 | - | 8 | 1326 | 9007335 | |
U29539.1 | - | 8 | 1148 | ||
U51239.1 | - | 8 | 1169 |