Gene Transcripts

Gene Symbol
Nras
Entrez Gene ID
18176
Gene Name
neuroblastoma ras oncogene
Gene Aliases
AV095280, N-ras
Chromosome Location
Chr. 3: 103058285 - 103067914 on Build GRCm38
UniGene ID
Mm.400954
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010937.2 NP_035067.2 2, 3 292
GenBank mRNA AK011704.1 - 2, 3 239
AK150357.1 - 2, 3 231
AK165960.1 - 2, 3 236
BC051443.1 - 2, 3 215 30061371
BC058755.1 - 2, 3 224 6475312
BY321840.1 - 2, 3 292
CT010359.1 - 1, 2 119
X13664.1 - 2, 3 336

View Full Product Details