Gene Transcripts

Gene Symbol
Nck2
Entrez Gene ID
17974
Gene Name
non-catalytic region of tyrosine kinase adaptor protein 2
Gene Aliases
4833426I10Rik, Grb4, NCKbeta, Nck-2
Chromosome Location
Chr. 1: 43445749 - 43570518 on Build GRCm38
UniGene ID
Mm.389903
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010879.3 NP_035009.3 4 1100
GenBank mRNA AF043260.1 - 3 737
AK014949.1 - 4 1094
AK051665.1 - 4 1100
BC011071.1 - 2 665 3673483
BC034255.1 - 4 840 3157491

View Full Product Details