Gene Transcripts

Gene Symbol
CREB3L1
Entrez Gene ID
90993
Gene Name
cAMP responsive element binding protein 3 like 1
Gene Aliases
OASIS
Chromosome Location
Chr. 11: 46277638 - 46321422 on Build GRCh38
UniGene ID
Hs.405961
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052854.3 NP_443086.1 6, 7 1369
XM_006718380.3 XP_006718443.1 6, 7 1346
GenBank mRNA AB063321.1 - 6, 7 990
AK075538.1 - 1, 2 140
AK314072.1 - 6, 7 1347
BC014097.1 - 6, 7 1012 4546950
BC015781.1 - 6, 7 998 4856946

View Full Product Details