Gene Transcripts

Gene Symbol
Pld3
Entrez Gene ID
18807
Gene Name
phospholipase D family, member 3
Gene Aliases
Sam-9
Chromosome Location
Chr. 7: 27531645 - 27553213 on Build GRCm38
UniGene ID
Mm.6483
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011116.1 NP_035246.1 7 698
GenBank mRNA AF026124.1 - 7 698
AK153508.1 - 5 387
BC068170.1 - 7 695
BC076586.1 - 7 762 30604186

View Full Product Details