Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011157.2 | NP_035287.1 | 1, 2 | 122 | |
GenBank mRNA | AK151357.1 | - | 1, 2 | 126 | |
AK157945.1 | - | 1, 2 | 123 | ||
AK168124.1 | - | 1, 2 | 125 | ||
BC037076.1 | - | 1, 2 | 112 | 5038568 | |
J04549.1 | - | 1, 2 | 119 | ||
M34603.1 | - | 1, 2 | 123 | ||
X16133.1 | - | 1, 2 | 122 |