Gene Transcripts

Gene Symbol
Psme1
Entrez Gene ID
19186
Gene Name
proteasome (prosome, macropain) activator subunit 1 (PA28 alpha)
Gene Aliases
AW413925, PA28a
Chromosome Location
Chr. 14: 55578494 - 55581527 on Build GRCm38
UniGene ID
Mm.830
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011189.1 NP_035319.1 3, 4 224
GenBank mRNA AK008514.1 - 3, 4 199
AK019390.1 - 3, 4 252
AK150708.1 - 3, 4 268
AK150975.1 - 3, 4 267
BC088981.1 - 3, 4 244 5689048
D87909.1 - 3, 4 224
U60328.1 - 3, 4 143

View Full Product Details