Gene Transcripts

Gene Symbol
Rbmx
Entrez Gene ID
19655
Gene Name
RNA binding motif protein, X chromosome
Gene Aliases
hnRNP G
Chromosome Location
Chr. X: 57383348 - 57393042 on Build GRCm38
UniGene ID
Mm.28275
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166623.1 NP_001160095.1 2 294
NM_011252.4 NP_035382.1 2 291
GenBank mRNA AJ237846.1 - 2 221
AJ237847.1 - 2 236
AK083899.1 - 2 291
AK135164.1 - 2 296
AK165006.1 - 2 237
AK207357.1 - 2 155
BC003710.1 - 2 289 3599678
BY279114.1 - 2 291
CK619886.1 - 1 97

View Full Product Details