Gene Transcripts

Gene Symbol
Pou2f3
Entrez Gene ID
18988
Gene Name
POU domain, class 2, transcription factor 3
Gene Aliases
Epoc-1, Oct-11a, Oct11, Otf-11, Otf11, Skin, Skin-1a, Skn-1a, Skn-li
Chromosome Location
Chr. 9: 43123927 - 43205755 on Build GRCm38
UniGene ID
Mm.436650
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011139.2 NP_035269.2 4 226
GenBank mRNA AK153697.1 - 316
L14677.1 - 4 224
Z18537.1 - 4 227

View Full Product Details