Gene Transcripts

Gene Symbol
Pou2f2
Entrez Gene ID
18987
Gene Name
POU domain, class 2, transcription factor 2
Gene Aliases
Oct-2, Oct2a, Oct2b, Otf-2, Otf2
Chromosome Location
Chr. 7: 25091114 - 25179992 on Build GRCm38
UniGene ID
Mm.76683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163554.1 NP_001157026.1 15 1526
NM_001163555.1 NP_001157027.1 14 1518
NM_001163556.1 NP_001157028.1 14 1452
NM_011138.2 NP_035268.2 14 1500
GenBank mRNA AK156322.1 - 14 1466
AY746973.1 - 14 1480
AY746974.1 - 14 1857
BC050258.1 - 14 1465
BC104488.1 - 14 1452 40046281
BC105647.1 - 14 1452 40046279
BC105920.1 - 14 1518 40046289
BC105921.1 - 14 1452 40046286
S55236.1 - 7 957
X53654.1 - 1734
X57089.1 - 7 957
X57936.1 - 14 1452
X57937.1 - 14 1500
X57938.1 - 14 1518
X57939.1 - 14 1316
X57940.1 - 15 1526
X57941.1 - 13 1335

View Full Product Details