Gene Transcripts

Gene Symbol
Rlim
Entrez Gene ID
19820
Gene Name
ring finger protein, LIM domain interacting
Gene Aliases
AL022832, AW743871, Ha1r, Rlim1, Rnf12
Chromosome Location
Chr. X: 103957164 - 103981297 on Build GRCm38
UniGene ID
Mm.427762
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011276.3 NP_035406.3 3 403
GenBank mRNA AF069992.1 - 3 358
AK013207.1 - 3 375
AK029295.1 - 3 397
BC012960.1 - 1 136 4193003

View Full Product Details