Gene Transcripts

Gene Symbol
Shbg
Entrez Gene ID
20415
Gene Name
sex hormone binding globulin
Gene Aliases
ABP
Chromosome Location
Chr. 11: 69610281 - 69618025 on Build GRCm38
UniGene ID
Mm.1431
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011367.2 NP_035497.1 3 297
GenBank mRNA AK010959.1 - 3 297
BC120785.1 - 3 282 40129708
BC120787.1 - 3 282 40129710
U85644.1 - 3 260

View Full Product Details