Gene Transcripts

Gene Symbol
Rgs9
Entrez Gene ID
19739
Gene Name
regulator of G-protein signaling 9
Gene Aliases
RGS9-1, Rgs9-2
Chromosome Location
Chr. 11: 109225348 - 109298181 on Build GRCm38
UniGene ID
Mm.38548
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165934.1 NP_001159406.1 8 705
NM_011268.2 NP_035398.2 8 705
GenBank mRNA AF011358.1 - 8 590
AF125046.1 - 8 661
AK085443.1 - 8 631
AK138159.1 - 8 615
AK164961.1 - 8 659
AK166518.1 - 549
BC129899.1 - 8 571 40111531

View Full Product Details