Gene Transcripts

Gene Symbol
Stau1
Entrez Gene ID
20853
Gene Name
staufen (RNA binding protein) homolog 1 (Drosophila)
Gene Aliases
5830401L18Rik, AW549911, C85792, Stau
Chromosome Location
Chr. 2: 166947549 - 166996299 on Build GRCm38
UniGene ID
Mm.73276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109905.2 NP_001103375.1 3 493
NM_001109906.2 NP_001103376.1 3 493
NM_001291160.1 NP_001278089.1 3 493
NM_011490.4 NP_035620.1 3 493
GenBank mRNA AF061942.1 - 3 493
AF395842.1 - 2 214
AK005002.2 - 3 434
AK139532.1 - 3 440
AK145826.1 - 3 460
AK151835.1 - 3 299
AK152269.1 - 3 305
AK152744.1 - 3 299
AK159602.1 - 3 410
AK159781.1 - 3 463
AK160979.1 - 3 434
AK167637.1 - 3 434
AK193147.1 - 100
AY391773.1 - 2 226
BC012959.1 - 3 457 4188394
BC082277.1 - 3 430

View Full Product Details