Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011778.2 | NP_035908.1 | 4, 5 | 538 | |
GenBank mRNA | AF143956.1 | - | 4, 5 | 545 | |
AK149639.1 | - | 4, 5 | 548 | ||
AK159329.1 | - | 2, 3 | 232 | ||
AK181268.1 | - | 2, 3 | 256 | ||
BC029326.1 | - | 4, 5 | 497 | ||
BC131944.1 | - | 4, 5 | 485 | 40130221 | |
BC132617.1 | - | 4, 5 | 485 | 40130894 |