Gene Transcripts

Gene Symbol
Slc26a4
Entrez Gene ID
23985
Gene Name
solute carrier family 26, member 4
Gene Aliases
Pds, pendrin
Chromosome Location
Chr. 12: 31519814 - 31560637 on Build GRCm38
UniGene ID
Mm.100187
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011867.3 NP_035997.1 9, 10 1372
GenBank mRNA AF167411.1 - 9, 10 1373
BC140995.1 - 8, 9 1220 9055607

View Full Product Details