Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011894.2 | NP_036024.2 | 4 | 492 | |
GenBank mRNA | AB016835.1 | - | 4 | 320 | |
AK043375.1 | - | 537 | |||
AK161427.1 | - | 3 | 474 | ||
AK165033.1 | - | 4 | 526 | ||
BC018237.1 | - | 4 | 524 | 3482670 | |
BC053741.1 | - | 4 | 492 | 6336511 |