Gene Transcripts
- Gene Symbol
- Mapk14
- Entrez Gene ID
- 26416
- Gene Name
- mitogen-activated protein kinase 14
- Gene Aliases
- CSBP2, Crk1, Csbp1, Mxi2, PRKM14, PRKM15, p38, p38-alpha, p38MAPK, p38a, p38alpha
- Chromosome Location
- Chr. 17: 28691342 - 28748405 on Build GRCm38
- UniGene ID
- Mm.311337
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001168508.1 |
NP_001161980.1
|
3, 4 |
697 |
|
NM_001168513.1 |
NP_001161985.1
|
3, 4 |
298 |
|
NM_001168514.1 |
NP_001161986.1
|
4, 5 |
742 |
|
NM_011951.3 |
NP_036081.1
|
3, 4 |
697 |
|
GenBank mRNA |
AF128892.1 |
- |
3, 4 |
595 |
|
AK089059.1 |
- |
3, 4 |
611 |
|
AK133684.1 |
- |
3, 4 |
610 |
|
AK151348.1 |
- |
3, 4 |
299 |
|
AK153025.1 |
- |
3, 4 |
299 |
|
BC012235.1 |
- |
3, 4 |
697 |
4195842 |
BC018227.1 |
- |
4, 5 |
639 |
|
BC085303.1 |
- |
3, 4 |
673 |
5389457 |
CX568915.1 |
- |
2, 3 |
91 |
|
D83073.1 |
- |
3, 4 |
528 |
|
U10871.1 |
- |
3, 4 |
595 |
|
View Full Product Details