Gene Transcripts

Gene Symbol
Vnn3
Entrez Gene ID
26464
Gene Name
vanin 3
Gene Aliases
Chromosome Location
Chr. 10: 23851462 - 23869843 on Build GRCm38
UniGene ID
Mm.27334
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011979.2 NP_036109.3 1, 2 343
GenBank mRNA AJ132103.1 - 1, 2 333
BC111521.2 - 1, 2 315 40047284

View Full Product Details