Gene Transcripts

Gene Symbol
Zw10
Entrez Gene ID
26951
Gene Name
zw10 kinetochore protein
Gene Aliases
6330566F14Rik, MmZw10
Chromosome Location
Chr. 9: 49055581 - 49078775 on Build GRCm38
UniGene ID
Mm.24791
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012039.2 NP_036169.1 8, 9 1187
GenBank mRNA AK167251.1 - 8, 9 1156
AK167937.1 - 8, 9 1116
AK168479.1 - 8, 9 1156
AK208616.1 - 2, 3 236
BC012435.1 - 8, 9 1136 3711269
BC067405.1 - 3226

View Full Product Details