Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083959.1 | NP_001077428.1 | 12 | 1535 | |
NM_001083960.1 | NP_001077429.1 | 12 | 1496 | ||
NM_012046.2 | NP_036176.1 | 13 | 1610 | ||
GenBank mRNA | AF126400.1 | - | 12 | 1546 | |
AF149309.1 | - | 12 | 1470 | ||
AF163053.1 | - | 12 | 1507 | ||
AF163054.1 | - | 12 | 1495 | ||
AF165313.1 | - | 12 | 1489 | ||
AF167439.1 | - | 12 | 1528 | ||
AF169386.1 | - | 13 | 1618 | ||
AF173848.1 | - | 13 | 1603 | ||
AV282800.1 | - | 1 | 143 | ||
BC064120.1 | - | 3 | 863 |