Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013654.3 | NP_038682.1 | 2 | 232 | |
GenBank mRNA | AF128193.1 | - | 2 | 197 | |
AF128194.1 | - | 2 | 197 | ||
AK078827.1 | - | 2 | 233 | ||
BC061126.1 | - | 2 | 230 | 30276350 | |
L04694.1 | - | 2 | 194 | ||
S50588.1 | - | 2 | 180 | ||
S71251.1 | - | 2 | 212 | ||
Z12297.1 | - | 2 | 180 |