Gene Transcripts
- Gene Symbol
- Plekhb1
- Entrez Gene ID
- 27276
- Gene Name
- pleckstrin homology domain containing, family B (evectins) member 1
- Gene Aliases
- AI315628, PHR1, Phret1, evt-1
- Chromosome Location
- Chr. 7: 100643896 - 100662394 on Build GRCm38
- UniGene ID
- Mm.491216
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF000272.1 |
- |
7 |
713 |
|
AF100613.1 |
- |
5 |
574 |
|
View Full Product Details