Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168672.1 | NP_001162143.1 | 4 | 409 | |
NM_013882.2 | NP_038910.1 | 3 | 379 | ||
GenBank mRNA | AJ222580.1 | - | 3 | 349 | |
AK012870.1 | - | 4 | 410 | ||
AK081582.1 | - | 4 | 372 | ||
BC027213.1 | - | 3 | 380 | 4039841 |