Gene Transcripts

Gene Symbol
Dnajc12
Entrez Gene ID
30045
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 12
Gene Aliases
Jdp1, mJDP1
Chromosome Location
Chr. 10: 63382443 - 63408840 on Build GRCm38
UniGene ID
Mm.32550
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253685.1 NP_001240614.1 3 189
NM_013888.3 NP_038916.1 3 326
GenBank mRNA AF132906.2 - 3 275
AK002649.1 - 1 14
AK032207.1 - 2 1695
BC013501.1 - 3 233 4222828
BC071211.1 - 2 1591
BF453706.1 - 3 189
DV042983.1 - 3 341

View Full Product Details