Gene Transcripts

Gene Symbol
Crlf2
Entrez Gene ID
57914
Gene Name
cytokine receptor-like factor 2
Gene Aliases
CRLM2, Ly114, Tpte2, Tslpr
Chromosome Location
Chr. 5: 109554709 - 109559017 on Build GRCm38
UniGene ID
Mm.35771
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164735.1 NP_001158207.1 8 1267
NM_016715.4 NP_057924.3 8 1234
GenBank mRNA AB031333.1 - 8 1182
AB039945.1 - 8 1130
AF201963.1 - 8 1318
AF232936.1 - 8 1198
AK010291.1 - 3 426
AK149928.1 - 8 1236
AK170466.1 - 5 1096
BC016580.1 - 8 1312 2655857
BC023788.1 - 8 1222 5346748

View Full Product Details