Gene Transcripts

Gene Symbol
PRKD3
Entrez Gene ID
23683
Gene Name
protein kinase D3
Gene Aliases
EPK2, PKC-NU, PKD3, PRKCN, nPKC-NU
Chromosome Location
Chr. 2: 37250502 - 37324803 on Build GRCh38
UniGene ID
Hs.660757
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005813.4 NP_005804.1 1 797
XM_005264237.3 XP_005264294.1 2 1070
XM_017003741.1 XP_016859230.1 2 1020
GenBank mRNA AB015982.2 - 1 797
BC012820.2 - 1 756
BC030706.2 - 1 742 5266769

View Full Product Details