Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016763.2 | NP_058043.3 | 6 | 849 | |
GenBank mRNA | AF233685.1 | - | 6 | 849 | |
AK002368.1 | - | 6 | 844 | ||
AK013340.1 | - | 6 | 845 | ||
AK077735.1 | - | 6 | 843 | ||
AK088685.1 | - | 6 | 865 | ||
AK179857.1 | - | 1 | 84 | ||
BC027517.1 | - | 6 | 849 | 3376098 | |
U96116.1 | - | 6 | 950 |