Gene Transcripts

Gene Symbol
Rab33b
Entrez Gene ID
19338
Gene Name
RAB33B, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 3: 51483966 - 51496228 on Build GRCm38
UniGene ID
Mm.1664
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016858.2 NP_058554.1 2 1974
GenBank mRNA AB004665.1 - 2 1924
AK004974.1 - 2 1710
AK087967.1 - 2 1715
AK139821.1 - 2 1960
AK144706.1 - 2 1714
AK159674.1 - 2 1711
AK163783.1 - 2 1961
AK170483.1 - 2 1603
BC065076.1 - 2 1974 6815992

View Full Product Details