Gene Transcripts
- Gene Symbol
- Bysl
- Entrez Gene ID
- 53414
- Gene Name
- bystin-like
- Gene Aliases
- Bys, Enp1
- Chromosome Location
- Chr. 17: 47599331 - 47611492 on Build GRCm38
- UniGene ID
- Mm.472101
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016859.3 |
NP_058555.3
|
4, 5 |
1030 |
|
GenBank mRNA |
AF007802.1 |
- |
2, 3 |
259 |
|
AK019239.1 |
- |
4, 5 |
765 |
|
AK032993.1 |
- |
4, 5 |
762 |
|
AK049195.1 |
- |
4, 5 |
1023 |
|
AK051704.1 |
- |
4, 5 |
1020 |
|
AK083033.1 |
- |
4, 5 |
1017 |
|
AK143027.1 |
- |
4, 5 |
1031 |
|
AK168301.1 |
- |
4, 5 |
768 |
|
BC017530.1 |
- |
4, 5 |
757 |
4925307 |
View Full Product Details