Gene Transcripts

Gene Symbol
Prkx
Entrez Gene ID
19108
Gene Name
protein kinase, X-linked
Gene Aliases
PKX1, Pkare
Chromosome Location
Chr. X: 77760972 - 77796288 on Build GRCm38
UniGene ID
Mm.290338
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016979.1 NP_058675.1 2 456
GenBank mRNA AJ238004.1 - 2 339
AK034681.1 - 2 775
AK036432.1 - 2 421
AK037141.1 - 2 450
AK039088.1 - 2 471
AK081548.1 - 2 436
AK139510.1 - 2 464
AK149937.1 - 2 434
AK150588.1 - 2 418
AK152166.1 - 2 253
AK154447.1 - 2 436
AK169322.1 - 2 407
BC006875.1 - 2 456 3596013

View Full Product Details