Gene Transcripts
- Gene Symbol
- Nckap1
- Entrez Gene ID
- 50884
- Gene Name
- NCK-associated protein 1
- Gene Aliases
- C79304, H19, Hem-2, Hem2, Nap1, mKIAA0587, mh19, p125Nap1
- Chromosome Location
- Chr. 2: 80500512 - 80581259 on Build GRCm38
- UniGene ID
- Mm.25203
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290745.1 |
NP_001277674.1
|
4, 5 |
555 |
|
NM_016965.3 |
NP_058661.1
|
3, 4 |
537 |
|
GenBank mRNA |
AJ534525.1 |
- |
3, 4 |
320 |
|
AK122318.1 |
- |
3, 4 |
407 |
|
AK191629.1 |
- |
3, 4 |
254 |
|
AK194160.1 |
- |
3, 4 |
254 |
|
AK198467.1 |
- |
3, 4 |
254 |
|
AK213144.1 |
- |
3, 4 |
254 |
|
BC060197.1 |
- |
3, 4 |
516 |
|
BC127067.1 |
- |
4, 5 |
337 |
|
BU055892.1 |
- |
3, 4 |
613 |
|
View Full Product Details