Gene Transcripts

Gene Symbol
Ggcx
Entrez Gene ID
56316
Gene Name
gamma-glutamyl carboxylase
Gene Aliases
Chromosome Location
Chr. 6: 72414296 - 72431106 on Build GRCm38
UniGene ID
Mm.19937
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019802.5 NP_062776.1 8, 9 1262
GenBank mRNA AF087938.1 - 8, 9 1237
AK033496.1 - 8, 9 1189
AK036577.1 - 8, 9 1236
AK135425.1 - 8, 9 1249
AK165982.1 - 8, 9 1237
BC138763.1 - 8, 9 1187 8861785
BC138764.1 - 8, 9 1187 8861786

View Full Product Details